missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Ethyl Ketone ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 78-93-3
Molecular Formula CH3COCH2CH3
Formula Weight 72.11
Chemical Name or Material Methyl Ethyl Ketone
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK62401 Avantor Performance
624006
Glass 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK624020 Avantor Performance
624019
Lined Steel Dr 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK62404 Avantor Performance
624008
Glass 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

78-93-3
72.11
ACS Reagent Grade
CH3COCH2CH3
Methyl Ethyl Ketone

Specifications

72.11
78-93-3
CH3COCH2CH3
ACS Reagent Grade
1L
Glass
Methyl Ethyl Ketone