Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl elaidate, 95%

$471.01 - $471.01

Chemical Identifiers

Chemical Name or Material Methyl elaidate
CAS 1937-62-8
Molecular Formula C19H36O2
Formula Weight 296.5
MDL Number MFCD00066521
Synonym Elaidic acid methyl ester; trans-9-Octadecenoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3045809

 
alfa aesar™
H3045809
10g Each for $471.01
Description & Specifications

Specifications

Formula Weight 296.5
Chemical Name or Material Methyl elaidate
TSCA TSCA
Assay 95%
Molecular Formula C19H36O2
MDL Number MFCD00066521
CAS 1937-62-8
Synonym Elaidic acid methyl ester; trans-9-Octadecenoic acid methyl ester
Melting Point 9° to 10°C
Quantity 10g