missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Methyl-DL-phenylalanine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444120010

 View more versions of this product

Catalog No. AC444120010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

α-Methyl-DL-phenylalanine
Glass Bottle
284°C to 286°C
1132-26-9
C10H13NO2
HYOWVAAEQCNGLE-UHFFFAOYSA-N
2-amino-2-methyl-3-phenylpropanoic acid
108055
Powder
Authentic
White
1g
97.5% min. (HPLC)
+/--2-amino-2-methyl-3-phenylpropionic acid, 2-amino-2-methyl-3-phenyl-propanoic acid, 2-amino-2-methyl-3-phenylpropionic acid, 2-methylphenylalanine #, alpha-methyl-dl-phenylalanine, alpha-methylphenylalanine, h-alpha-me-dl-phe-oh, h-dl-me phe-oh, h-me-dl-phe-oh, phenylalanine, alpha-methyl
CC(CC1=CC=CC=C1)(C(=O)O)N
179.22
179.22
98%

Chemical Identifiers

1132-26-9
179.22
+/--2-amino-2-methyl-3-phenylpropionic acid, 2-amino-2-methyl-3-phenyl-propanoic acid, 2-amino-2-methyl-3-phenylpropionic acid, 2-methylphenylalanine #, alpha-methyl-dl-phenylalanine, alpha-methylphenylalanine, h-alpha-me-dl-phe-oh, h-dl-me phe-oh, h-me-dl-phe-oh, phenylalanine, alpha-methyl
2-amino-2-methyl-3-phenylpropanoic acid
C10H13NO2
HYOWVAAEQCNGLE-UHFFFAOYSA-N
108055
CC(CC1=CC=CC=C1)(C(=O)O)N
SDS Documents