Please login to your online account to display your discounted pricing

α-Methyl-DL-phenylalanine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material α-Methyl-DL-phenylalanine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H13NO2
Formula Weight 179.22
Synonym (+/-)-2-Amino-2-methyl-3-phenylpropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444120010

  SDS
thermo fisher scientific
AC444120010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444120050

 
thermo fisher scientific
AC444120050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 179.22
Quantity 1g
Color White
Chemical Name or Material α-Methyl-DL-phenylalanine
Assay Percent Range 98%
Molecular Formula C10H13NO2
CAS 1132-26-9
Synonym (+/-)-2-Amino-2-methyl-3-phenylpropionic acid
Infrared Spectrum Authentic