missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 4358-87-6 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.176 |
MDL Number | MFCD00004493 |
InChI Key | ITATYELQCJRCCK-UHFFFAOYSA-N |
Synonym | methyl mandelate, methyl dl-mandelate, mandelic acid, methyl ester, methyl phenylglycolate, methyl hydroxy phenyl acetate, +/--methyl mandelate, mandelic acid methyl ester, dl-mandelic acid methyl ester, dl-mandelic acid, methyl ester, methyl 2-hydroxy-2-phenyl-acetate |
PubChem CID | 78066 |
IUPAC Name | methyl 2-hydroxy-2-phenylacetate |
SMILES | COC(=O)C(C1=CC=CC=C1)O |
Chemical Identifiers
4358-87-6 | |
166.176 | |
ITATYELQCJRCCK-UHFFFAOYSA-N | |
78066 | |
COC(=O)C(C1=CC=CC=C1)O |
C9H10O3 | |
MFCD00004493 | |
methyl mandelate, methyl dl-mandelate, mandelic acid, methyl ester, methyl phenylglycolate, methyl hydroxy phenyl acetate, +/--methyl mandelate, mandelic acid methyl ester, dl-mandelic acid methyl ester, dl-mandelic acid, methyl ester, methyl 2-hydroxy-2-phenyl-acetate | |
methyl 2-hydroxy-2-phenylacetate |
Specifications
250°C | |
56°C | |
4358-87-6 | |
MFCD00004493 | |
ITATYELQCJRCCK-UHFFFAOYSA-N | |
methyl 2-hydroxy-2-phenylacetate | |
78066 | |
Crystal-Powder at 20°C | |
Methyl DL-Mandelate |
White | |
25g | |
C9H10O3 | |
methyl mandelate, methyl dl-mandelate, mandelic acid, methyl ester, methyl phenylglycolate, methyl hydroxy phenyl acetate, +/--methyl mandelate, mandelic acid methyl ester, dl-mandelic acid methyl ester, dl-mandelic acid, methyl ester, methyl 2-hydroxy-2-phenyl-acetate | |
COC(=O)C(C1=CC=CC=C1)O | |
166.176 | |
166.18 | |
≥98.0% (GC) |
Safety and Handling
EINECSNumber : (3)-3842
TSCA : Yes