Learn More
Methyl DL-lactate, 99%, Thermo Scientific Chemicals
$124.84 - $412.52
Chemical Identifiers
CAS | 547-64-8 |
---|---|
Molecular Formula | C4H8O3 |
Molecular Weight (g/mol) | 104.105 |
MDL Number | MFCD00066367 |
InChI Key | LPEKGGXMPWTOCB-UHFFFAOYSA-N |
Synonym | methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate |
PubChem CID | 11040 |
ChEBI | CHEBI:83221 |
IUPAC Name | methyl 2-hydroxypropanoate |
SMILES | CC(C(=O)OC)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH60455AA
|
Thermo Scientific Chemicals
H60455AA |
5 mL |
Each for $124.84
|
|
|||||
AAH60455AC
|
Thermo Scientific Chemicals
H60455AC |
25 mL |
Each for $412.52
|
|
|||||
Description
Methyl DL-lactate is used as a solvent for nitrocellulose, cellulase acetate, cellulase acetobutyrote and cellulase acetaprapionate. It is used in the manufacture of lacquers and dopes where it contributes high tolerance for diluents, good flaw and blush resistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl DL-lactate is used as a solvent for nitrocellulose, cellulase acetate, cellulase acetobutyrote and cellulase acetaprapionate. It is used in the manufacture of lacquers and dopes where it contributes high tolerance for diluents, good flaw and blush resistance.
Solubility
Decomposes in water.
Notes
Moisture Sensitive. Store away from moisture and strong bases. Do not store together with acids. Store under dry inert gas. Incompatible with oxidizing agents.
Chemical Identifiers
547-64-8 | |
104.105 | |
LPEKGGXMPWTOCB-UHFFFAOYSA-N | |
11040 | |
methyl 2-hydroxypropanoate |
C4H8O3 | |
MFCD00066367 | |
methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate | |
CHEBI:83221 | |
CC(C(=O)OC)O |
Specifications
547-64-8 | |
51°C (124°F) | |
1.413 | |
5 mL | |
1720586 | |
methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate | |
LPEKGGXMPWTOCB-UHFFFAOYSA-N | |
methyl 2-hydroxypropanoate | |
11040 | |
104.1 | |
Methyl DL-lactate |
1.093 | |
C4H8O3 | |
MFCD00066367 | |
UN3272 | |
Moisture sensitive | |
Decomposes in water. | |
CC(C(=O)OC)O | |
104.105 | |
CHEBI:83221 | |
99% |
Safety and Handling
GHS H Statement
H226-H319-H335
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H226-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 208-930-0
RTECSNumber : OD5670000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only