missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl DL-2-Methylbutyrate 98.0+%, TCI America™

Manufacturer:  TCI America M0758100ML

 View more versions of this product

Catalog No. M0758100ML



Specifications

Specifications

Methyl DL-2-Methylbutyrate
Colorless
868-57-5
MFCD00009335
methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
CCC(C)C(=O)OC
116.16
116.16
≥98.0% (GC)
115°C
100mL
C6H12O2
3272
OCWLYWIFNDCWRZ-UHFFFAOYSA-N
methyl 2-methylbutanoate
13357
Clear Liquid at 20°C

Chemical Identifiers

868-57-5
116.16
OCWLYWIFNDCWRZ-UHFFFAOYSA-N
13357
CCC(C)C(=O)OC
C6H12O2
MFCD00009335
methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
methyl 2-methylbutanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents