Learn More
Methyl DL-2-bromopropionate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 173395000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl DL-2-bromopropionate | |
5445-17-0 | |
143.0°C to 145.0°C (19.0mmHg) | |
Authentic | |
Glass bottle | |
1.4509 to 1.4530 | |
MFCD00000143 | |
02, 253 | |
methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester | |
ACEONLNNWKIPTM-UHFFFAOYSA-N | |
methyl 2-bromopropanoate | |
95576 | |
99% |
99% | |
1.4970g/mL | |
51°C | |
98.5% min. (GC) | |
C4H7BrO2 | |
CH3CH(Br)CO2CH3 | |
500g | |
1.497 | |
Solubility in water: immiscible | |
CC(C(=O)OC)Br | |
167 | |
167 |
Chemical Identifiers
5445-17-0 | |
167 | |
ACEONLNNWKIPTM-UHFFFAOYSA-N | |
95576 | |
CC(C(=O)OC)Br |
C4H7BrO2 | |
MFCD00000143 | |
methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester | |
methyl 2-bromopropanoate |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 226-642-3
TSCA : TSCA
RUO â Research Use Only