Alfa Aesar™ Methyl diphenylphosphite
Manufacturer:  Alfa Aesar™ 3209703
 View more versions of this product
Catalog No. AA3209703
Specifications
Specifications
Methyl diphenylphosphite
1g
C13H13O3P
UN3278
NYCZNDFWFCCTPA-UHFFFAOYSA-N
methyl diphenyl phosphite
520764
Liquid
Hygroscopic
3577-87-5
MFCD00014886
diphenyl methyl phosphite, methyldiphenylphosphite, methoxydiphenoxyphosphine, methoxy-diphenoxy-phosphane, methyl diphenylphosphite 1g, phosphorous acid, methyl diphenyl ester
COP(OC1=CC=CC=C1)OC2=CC=CC=C2
248.218
248.21
Chemical Identifiers
3577-87-5
248.218
NYCZNDFWFCCTPA-UHFFFAOYSA-N
520764
COP(OC1=CC=CC=C1)OC2=CC=CC=C2
C13H13O3P
MFCD00014886
diphenyl methyl phosphite, methyldiphenylphosphite, methoxydiphenoxyphosphine, methoxy-diphenoxy-phosphane, methyl diphenylphosphite 1g, phosphorous acid, methyl diphenyl ester
methyl diphenyl phosphite
Safety and Handling
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
SDS Documents