missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl diphenylphosphinite, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1155514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl diphenylphosphinite | |
1.078 | |
>110°C (230°F) | |
1.604 | |
25 g | |
Air and moisture sensitive | |
OAADXJFIBNEPLY-UHFFFAOYSA-N | |
methoxy(diphenyl)phosphane | |
77636 | |
≥98% |
4020-99-9 | |
148°C to 150°C (6 mmHg) | |
C13H13OP | |
MFCD00048025 | |
1640480 | |
methoxydiphenylphosphine, methyl diphenylphosphinite, diphenylphosphinous acid methyl ester, phosphinous acid, diphenyl-, methyl ester, methoxy diphenyl phosphane, pubchem6470, acmc-209jbw, diphenyl methoxy phosphine | |
COP(C1=CC=CC=C1)C1=CC=CC=C1 | |
216.22 | |
216.22 |
Chemical Identifiers
4020-99-9 | |
216.22 | |
OAADXJFIBNEPLY-UHFFFAOYSA-N | |
77636 | |
COP(C1=CC=CC=C1)C1=CC=CC=C1 |
C13H13OP | |
MFCD00048025 | |
methoxydiphenylphosphine, methyl diphenylphosphinite, diphenylphosphinous acid methyl ester, phosphinous acid, diphenyl-, methyl ester, methoxy diphenyl phosphane, pubchem6470, acmc-209jbw, diphenyl methoxy phosphine | |
methoxy(diphenyl)phosphane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 223-683-9
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only