missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl dimethoxyacetate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 414180250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl dimethoxyacetate | |
89-91-8 | |
160°C to 163°C | |
Authentic | |
Glass bottle | |
1.4040 to 1.406 | |
MFCD00008484 | |
1.09 | |
NZTCVGHPDWAALP-UHFFFAOYSA-N | |
methyl 2,2-dimethoxyacetate | |
66647 | |
98% |
99% | |
1.0900g/mL | |
63°C | |
97.5% min. (GC) | |
C5H10O4 | |
(CH3O)2CHCOOCH3 | |
25g | |
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
COC(C(=O)OC)OC | |
134.13 | |
134.13 |
Chemical Identifiers
89-91-8 | |
134.13 | |
NZTCVGHPDWAALP-UHFFFAOYSA-N | |
66647 | |
COC(C(=O)OC)OC |
C5H10O4 | |
MFCD00008484 | |
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
methyl 2,2-dimethoxyacetate |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 201-950-0
TSCA : TSCA
RUO â Research Use Only