missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl dimethoxyacetate, 97+%

Manufacturer:  Alfa Aesar™ A1217106

 View more versions of this product

Catalog No. AAA1217106



Specifications

Specifications

Methyl dimethoxyacetate
≥97%
MFCD00008484
Dimethoxyacetic acid methyl ester; Glyoxylic acid dimethyl acetal methyl ester
COC(C(=O)OC)OC
134.131
160°C to 163°C
5g
89-91-8
C5H10O4
1757582
NZTCVGHPDWAALP-UHFFFAOYSA-N
methyl 2,2-dimethoxyacetate
66647
134.13
63°C (145°F)

Chemical Identifiers

89-91-8
134.131
NZTCVGHPDWAALP-UHFFFAOYSA-N
66647
COC(C(=O)OC)OC
C5H10O4
MFCD00008484
Dimethoxyacetic acid methyl ester; Glyoxylic acid dimethyl acetal methyl ester
methyl 2,2-dimethoxyacetate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents