Please login to your online account to display your discounted pricing

Methyl α-D-mannopyranoside, Fisher BioReagents


Quantity 25g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP253025

 View more versions of this product

Catalog No. BP253025

  • / Each
  • / Case of 8 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 194.2
Melting Point 193° to 196°C
Physical Form Powder/Solid
Quantity 25g
Molecular Formula C7H14O6
Color White
Identification Pass Test
FTIR Conforms to reference
Chemical Name or Material Methyl α-D-Mannopyranoside
ChemAlert Storage Symbol Gray
Assay Percent Range 99%
Purity approx. 99%
Specific Rotation 76 to 85° (+ or -)
Loss on Drying < 0.5%
Packaging Amber Glass
Synonym α-Methyl-D-mannoside
Recommended Storage RT
CAS 617-04-9

Specifications

Chemical Name or Material Methyl α-D-Mannopyranoside
CAS 617-04-9
Physical Form Powder/Solid
Assay Percent Range 99%
Molecular Formula C7H14O6
Formula Weight 194.2
Synonym α-Methyl-D-mannoside

  • Methyl α-D-mannopyranoside is a lectin inhibitory sugar
  • Competitive inhibitor of the binding of mannose by E. coli
  • Intended for research use only

Accessories
 

RT

SDS & Safety Documents