missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl α-D-mannopyranoside, ≥99%, Fisher BioReagents™
Supplier: Fisher BioReagents BP253010
Description
Methyl α-D-mannopyranoside, ≥99%
- Methyl α-D-mannopyranoside is a lectin inhibitory sugar
- Competitive inhibitor of the binding of mannose by E. coli
- Intended for research use only
Specifications
Methyl α-D-Mannopyranoside | |
193°C to 196°C | |
< 0.5% | |
Amber Glass | |
MFCD00063262 | |
76 to 85° (+ or -) | |
HOVAGTYPODGVJG-UHFFFAOYNA-N | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
101798 | |
194.2 | |
Powder or Solid |
617-04-9 | |
White | |
≥99% | |
C7H14O6 | |
10 g | |
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
COC1OC(CO)C(O)C(O)C1O | |
194.18 | |
CHEBI:43943 | |
≥99% |
Chemical Identifiers
617-04-9 | |
194.18 | |
HOVAGTYPODGVJG-UHFFFAOYNA-N | |
101798 | |
COC1OC(CO)C(O)C(O)C1O |
C7H14O6 | |
MFCD00063262 | |
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
CHEBI:43943 |
Safety and Handling
Recommended Storage : RT