missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl-β-D-glucopyranoside hemihydrate, 99%, ACROS Organics™

$327.47 - $327.47

Chemical Identifiers

CAS 7000-27-3
Assay Percent Range 99%
Molecular Formula 0·5 H2O
Formula Weight 203.19
MDL Number MFCD00149442
Chemical Name or Material Methyl-β-D-glucopyranoside hemihydrate, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC270920250 SDS Thermo Fisher Scientific
270920250
Glass bottle 25g Each for $327.47
Specifications Description & Specifications

Chemical Identifiers

7000-27-3
0·5 H2O
MFCD00149442
≥98.5%
99%
203.19
Methyl-β-D-glucopyranoside hemihydrate, 99%

Specifications

−34.2° (20°C c=1,water)
25g
(10% aq. soln.) clear, colorless
Glass bottle
Crystalline Powder
0·5 H2O
203.19
Methyl-β-D-glucopyranoside hemihydrate, 99%
White
7000-27-3
Authentic
-31° to -35° (20°C, 589nm) (c=4, water)
99%
5.4% max. (K.F.)
MFCD00149442
≥98.5%