missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl-beta-D-glucopyranoside hemihydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 270920250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl-β-D-glucopyranoside hemihydrate | |
7000-27-3 | |
White | |
98.5% min. (ELSD) (HPLC) | |
0·5 H2O | |
25 g | |
-34.2 | |
(10% in water) Clear colorless | |
COC1C(C(C(C(O1)CO)O)O)O.O | |
203.19 | |
203.19 | |
Crystalline Powder |
99% | |
111°C to 113°C | |
Authentic | |
Glass bottle | |
MFCD00149442 | |
−34.2° (20°C c=1,water) | |
methyl beta-d-glucoside, methyl beta-d-glucopyranoside, beta-methylglucoside, o1-methyl-glucose, beta-methyl-d-glucoside, methyl-beta-d-glucopyranoside, 1-o-methyl-beta-d-glucopyranoside, beta-d-methylglucopyranoside, methyl-beta-d-glucopyranoside hemihydrate, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol | |
QFDRIBVAVJNJDT-DEOAAGHXSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol;hydrate | |
75487602 | |
99% |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only