Please login to your online account to display your discounted pricing

Methyl-β-D-galactopyranoside, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC314980250

 View more versions of this product

Catalog No. AC314980250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 194.18
Quantity 25g
Color White
Specific Rotation Condition −18° (20° (c=1.5,MeOH))
Chemical Name or Material Methyl-β-D-galactopyranoside
Specific Rotation -16° to -20° (20°C, 589nm) (c=1.5, MeOH)
Water 1% max. (K.F.)
Assay Percent Range 98%
Molecular Formula C7H14O6
MDL Number MFCD00064357
CAS 1824-94-8
Packaging Glass bottle
Appearance of Solution (1% soln. in MeOH) clear, colorless
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Methyl-β-D-galactopyranoside
CAS 1824-94-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H14O6
Formula Weight 194.18
MDL Number MFCD00064357