Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl cyclopentene-3-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Methyl cyclopentene-3-carboxylate
CAS 58101-60-3
Formula Weight 126.15
MDL Number MFCD04038661
UN Number UN1993
Synonym Cyclopentene-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6674303

 
alfa aesar™
H6674303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6674306

 
alfa aesar™
H6674306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.15
Chemical Name or Material Methyl cyclopentene-3-carboxylate
Synonym Cyclopentene-3-carboxylic acid methyl ester
Health Hazard 1 Danger
Refractive Index 1.4500
Physical Form Liquid
Assay 97%
MDL Number MFCD04038661
CAS 58101-60-3
UN Number UN1993
Health Hazard 2 GHS H Statement
H318-H226-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density 1.03g/mL
Color Colorless to pale brown
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H226-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.