missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl cyclohexanecarboxylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 188820050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl cyclohexanecarboxylate | |
4630-82-4 | |
0.9950g/mL | |
60°C | |
98% | |
C8H14O2 | |
C6H11CO2CH3 | |
5 g | |
cyclohexanecarboxylic acid, methyl ester, methyl cyclohexanoate, methyl cyclohexylcarboxylate, cyclohexanecarboxylic acid methyl ester, hexahydrobenzoic acid methyl ester, methyl cyclohexylformate, methyl hexahydrobenzoate, methyl cyclohexane carboxylate, fema no. 3568, methyl ester of cyclohexanecarboxylic acid | |
ZQWPRMPSCMSAJU-UHFFFAOYSA-N | |
methyl cyclohexanecarboxylate | |
20748 | |
98% |
98% | |
Colorless to Yellow | |
183°C | |
Authentic | |
Glass bottle | |
1.4420 to 1.444 | |
MFCD00001458 | |
0.995 | |
Solubility in water: almost insoluble in water | |
COC(=O)C1CCCCC1 | |
142.20 | |
142.2 | |
Liquid |
Chemical Identifiers
4630-82-4 | |
142.20 | |
ZQWPRMPSCMSAJU-UHFFFAOYSA-N | |
20748 | |
COC(=O)C1CCCCC1 |
C8H14O2 | |
MFCD00001458 | |
cyclohexanecarboxylic acid, methyl ester, methyl cyclohexanoate, methyl cyclohexylcarboxylate, cyclohexanecarboxylic acid methyl ester, hexahydrobenzoic acid methyl ester, methyl cyclohexylformate, methyl hexahydrobenzoate, methyl cyclohexane carboxylate, fema no. 3568, methyl ester of cyclohexanecarboxylic acid | |
methyl cyclohexanecarboxylate |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 225-050-2
RTECSNumber : GU8599000
RUO – Research Use Only