missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl cyanoacetate, 99%

Manufacturer:  Alfa Aesar™ A1555536

 View more versions of this product

Catalog No. AAA1555536



Description

Description

Applications
Methyl cyanoacetate is an intermediate in the preparation of α-cyanoacrylate, malononitrile, 1,1-cyclohexanediacetic acid dye. It is used as UV absorber or stabiliser, intermediate for pharmaceuticals. It may be used in the synthesis of various 1,2,5-tricarbonyl compounds.

Solubility
Soluble in water 117 g/L.

Notes
Light sensitive. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Methyl cyanoacetate
Mild
-23°C
105-34-0
MFCD00001939
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
COC(=O)CC#N
99.089
CHEBI:51928
99%
110°C (230°F)
204°C
500g
C4H5NO2
773945
ANGDWNBGPBMQHW-UHFFFAOYSA-N
methyl 2-cyanoacetate
7747
99.09

Chemical Identifiers

105-34-0
99.089
ANGDWNBGPBMQHW-UHFFFAOYSA-N
7747
methyl 2-cyanoacetate
C4H5NO2
MFCD00001939
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
CHEBI:51928
COC(=O)CC#N
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : AG4375000

TSCA : TSCA

Documents