Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl coumalate, 98%

$63.14 - $230.54

Chemical Identifiers

Chemical Name or Material Methyl coumalate
CAS 6018-41-3
Molecular Formula C7H6O4
Formula Weight 154.12
MDL Number MFCD00010120
Merck Index 14,2557
Synonym Coumalic acid methyl ester; Methyl 2-oxo-2H-pyran-5-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2533004

  SDS
alfa aesar™
B2533004
2g Each for $63.14

AAB2533009

  SDS
alfa aesar™
B2533009
10g Each for $230.54
Description & Specifications

Specifications

Formula Weight 154.12
Chemical Name or Material Methyl coumalate
Assay 98%
Molecular Formula C7H6O4
MDL Number MFCD00010120
CAS 6018-41-3
Merck Index 14,2557
Synonym Coumalic acid methyl ester; Methyl 2-oxo-2H-pyran-5-carboxylate
Boiling Point 178° to 180°C (60mmHg)
Melting Point 68° to 72°C
Quantity 2g

Applications
Methyl coumalate is a 2-pyrone and acts as dienophile in Diels-Alder reaction. It reacts with 1,3-butadienes at 100°C to yield tetrahydrocoumarins and 4-methoxycarbonyltricyclo[3.2.1.02,7]octenes. It also reacts with cyclohexadiene to afford tetrahydronaphthalene-2-carboxylate. It undergoes Diels-Alder reaction with unactivated alkenes to afford para-substituted adducts. It has been used as reagent in phosphine-catalyzed [4+3] annulation of modified allylic carbonates. It was also used in the preparation of 7-carboxyquinolizinium derivatives.

Solubility
Freely soluble in dichloromethane.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.