missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl cis-6-Octadecenoate, TCI America™

$32.08 - $108.47

Chemical Identifiers

CAS 2777-58-4
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00056193
InChI Key QRTVDKVXAFJVRU-YPKPFQOOSA-N
Synonym methyl petroselinate, methyl cis-6-octadecenoate, petroselinic acid methyl ester, unii-774w29lwkq, methyl z-octadec-6-enoate, methyl 6z-octadec-6-enoate, 6-octadecenoic acid, methyl ester, z, methyl z-6-octadecenoate, methyl 6z-6-octadecenoate #, cis-6-octadecenoic acid methyl ester
PubChem CID 5362717
IUPAC Name methyl (Z)-octadec-6-enoate
SMILES CCCCCCCCCCCC=CCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2306500MG View Documents TCI America
M2306500MG
500mg Each for $108.47
 
M2306100MG View Documents TCI America
M2306100MG
100mg Each for $32.08
 
Specifications

Chemical Identifiers

2777-58-4
296.495
QRTVDKVXAFJVRU-YPKPFQOOSA-N
5362717
CCCCCCCCCCCC=CCCCCC(=O)OC
C19H36O2
MFCD00056193
methyl petroselinate, methyl cis-6-octadecenoate, petroselinic acid methyl ester, unii-774w29lwkq, methyl z-octadec-6-enoate, methyl 6z-octadec-6-enoate, 6-octadecenoic acid, methyl ester, z, methyl z-6-octadecenoate, methyl 6z-6-octadecenoate #, cis-6-octadecenoic acid methyl ester
methyl (Z)-octadec-6-enoate

Specifications

160°C
-1°C
2777-58-4
MFCD00056193
QRTVDKVXAFJVRU-YPKPFQOOSA-N
methyl (Z)-octadec-6-enoate
5362717
Clear Liquid at 20°C
Methyl cis-6-Octadecenoate
Colorless
500mg
C19H36O2
methyl petroselinate, methyl cis-6-octadecenoate, petroselinic acid methyl ester, unii-774w29lwkq, methyl z-octadec-6-enoate, methyl 6z-octadec-6-enoate, 6-octadecenoic acid, methyl ester, z, methyl z-6-octadecenoate, methyl 6z-6-octadecenoate #, cis-6-octadecenoic acid methyl ester
CCCCCCCCCCCC=CCCCCC(=O)OC
296.495
296.50
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : No

Recommended Storage : Freezer