missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl cis-4-Aminocyclohexanecarboxylate Hydrochloride, TCI America™

$93.20 - $322.38

Chemical Identifiers

CAS 61367-16-6
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.671
MDL Number MFCD08274537
InChI Key NHAYDXCUCXRAMF-UHFFFAOYSA-N
Synonym cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
PubChem CID 18521575
IUPAC Name methyl 4-aminocyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19855G View Documents TCI America
M19855G
5g Each for $322.38
 
M19851G View Documents TCI America
M19851G
1g Each for $93.20
 
Specifications

Chemical Identifiers

61367-16-6
193.671
NHAYDXCUCXRAMF-UHFFFAOYSA-N
18521575
COC(=O)C1CCC(CC1)N.Cl
C8H16ClNO2
MFCD08274537
cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
methyl 4-aminocyclohexane-1-carboxylate;hydrochloride

Specifications

White
5g
C8H16ClNO2
cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
COC(=O)C1CCC(CC1)N.Cl
193.671
193.67
≥98.0% (N,T)
188°C
61367-16-6
MFCD08274537
NHAYDXCUCXRAMF-UHFFFAOYSA-N
methyl 4-aminocyclohexane-1-carboxylate;hydrochloride
18521575
Crystal-Powder at 20°C
Methyl cis-4-Aminocyclohexanecarboxylate Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No