missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Cinnamate 99.0+%, TCI America™
$33.56 - $186.41
Chemical Identifiers
| CAS | 103-26-4 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.188 |
| MDL Number | MFCD00008458 |
| InChI Key | CCRCUPLGCSFEDV-BQYQJAHWSA-N |
| Synonym | methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate |
| PubChem CID | 637520 |
| ChEBI | CHEBI:6857 |
| IUPAC Name | methyl (E)-3-phenylprop-2-enoate |
| SMILES | COC(=O)C=CC1=CC=CC=C1 |
Chemical Identifiers
| 103-26-4 | |
| 162.188 | |
| CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
| 637520 | |
| methyl (E)-3-phenylprop-2-enoate |
| C10H10O2 | |
| MFCD00008458 | |
| methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
| CHEBI:6857 | |
| COC(=O)C=CC1=CC=CC=C1 |
Specifications
| 103-26-4 | |
| 262°C | |
| MFCD00008458 | |
| methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
| COC(=O)C=CC1=CC=CC=C1 | |
| 162.188 | |
| CHEBI:6857 | |
| ≥99.0% (GC) | |
| Methyl Cinnamate |
| White | |
| C10H10O2 | |
| 25 g | |
| CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
| methyl (E)-3-phenylprop-2-enoate | |
| 637520 | |
| 162.19 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (3)-1750
RTECSNumber : GE0190000
TSCA : Yes