missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl cinnamate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 279760050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl cinnamate | |
103-26-4 | |
33.5°C to 36°C | |
260°C to 262°C | |
Authentic | |
Glass bottle | |
C6H5CH=CHCO2CH3 | |
5 g | |
15, 2299 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,glycerol,most fixed,oils and mineral oil | |
COC(=O)C=CC1=CC=CC=C1 | |
162.19 | |
CHEBI:6857 | |
98% |
98% | |
1mg KOH/g max. | |
White to Yellow | |
>110°C | |
97.5% min. (GC) | |
C10H10O2 | |
MFCD00008458 | |
09, 581 | |
methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
methyl (E)-3-phenylprop-2-enoate | |
637520 | |
162.19 | |
Fused Crystalline Mass |
Chemical Identifiers
103-26-4 | |
162.19 | |
CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
637520 | |
methyl (E)-3-phenylprop-2-enoate |
C10H10O2 | |
MFCD00008458 | |
methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
CHEBI:6857 | |
COC(=O)C=CC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 203-093-8
RTECSNumber : GE0190000
TSCA : TSCA
RUO – Research Use Only