Please login to your online account to display your discounted pricing

Methyl Cellulose, viscosity of 2% aqueous solution (25°C) 4,000cps, MP Biomedicals


Quantity 250g

Manufacturer: mp biomedicals  0215549683

 View more versions of this product

Catalog No. ICN15549683

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 250g
Chemical Name or Material Methyl Cellulose
Name Note Viscosity of 2% aqueous solution at 25°C: 4, 000cps
CAS 9004-67-5
Synonym Cellulose methyl ether
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Methyl Cellulose
Name Note Viscosity of 2% aqueous solution at 25°C: 4, 000cps
CAS 9004-67-5
Synonym Cellulose methyl ether
Accessories

Room Temperature