missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl carbamate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 269310050
Specifications
Methyl carbamate | |
Authentic | |
176.0°C to 177.0°C | |
54.0°C to 56.0°C | |
99% | |
99% | |
H2NCO2CH3 | |
03, 21 | |
carbamic acid methyl ester, carbamic acid, methyl ester, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech, methylkarbamat, methylkarbamat czech, methylurethan, unii-9wfx634x2t, urethylane | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
methyl carbamate | |
11722 | |
75.06 | |
99% |
93°C | |
Glass Bottle | |
White | |
5g | |
598-55-0 | |
C2H5NO2 | |
MFCD00007964 | |
15, 6110 | |
Solubility in water: 700g/L (20°C). Other solubilities: very soluble in ethanol,soluble in acetone and ether | |
COC(=O)N | |
75.06 | |
CHEBI:76606 | |
Adhering Crystals |
Chemical Identifiers
598-55-0 | |
75.06 | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
11722 | |
methyl carbamate |
C2H5NO2 | |
MFCD00007964 | |
carbamic acid methyl ester, carbamic acid, methyl ester, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech, methylkarbamat, methylkarbamat czech, methylurethan, unii-9wfx634x2t, urethylane | |
CHEBI:76606 | |
COC(=O)N |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 209-939-2
RTECSNumber : FC2450000
TSCA : TSCA