Learn More
Methyl carbamate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 269312500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
54.0°C to 56.0°C | |
99% | |
176.0°C to 177.0°C | |
99% | |
Authentic | |
MFCD00007964 | |
15, 6110 | |
urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
methyl carbamate | |
11722 | |
75.06 | |
250 g | |
Adhering Crystals |
Methyl carbamate | |
598-55-0 | |
White | |
C2H5NO2 | |
H2NCO2CH3 | |
03, 21 | |
Plastic Bottle | |
Solubility in water: 700g/L (20°C). Other solubilities: very soluble in ethanol,soluble in acetone and ether | |
COC(N)=O | |
75.07 | |
CHEBI:76606 | |
99% | |
93°C |
Chemical Identifiers
598-55-0 | |
75.07 | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
11722 | |
methyl carbamate |
C2H5NO2 | |
MFCD00007964 | |
urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
CHEBI:76606 | |
COC(N)=O |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 209-939-2
RTECSNumber : FC2450000
TSCA : TSCA
RUO – Research Use Only