Learn More
Methyl carbamate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1652322
Description
Methyl carbamate was used in the synthesis of protected aminocyclopropanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl carbamate was used in the synthesis of protected aminocyclopropanes.
Solubility
Soluble in water(20g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, metals, alkali metals.
Specifications
54°C to 56°C | |
598-55-0 | |
C2H5NO2 | |
635779 | |
urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
methyl carbamate | |
11722 | |
75.07 | |
1.14 | |
>100°C (212°F) |
Methyl carbamate | |
176°C to 178°C | |
MFCD00007964 | |
14,6036 | |
Soluble in water(20g/L). | |
COC(N)=O | |
75.07 | |
CHEBI:76606 | |
99% | |
100 g |
Chemical Identifiers
598-55-0 | |
75.07 | |
GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
11722 | |
methyl carbamate |
C2H5NO2 | |
MFCD00007964 | |
urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
CHEBI:76606 | |
COC(N)=O |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P332+P313-P362-P501c
H315-H319-H335-H351
EINECSNumber : 209-939-2
RTECSNumber : FC2450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only