missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl caprate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 385550250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl caprate | |
-18°C | |
0.8710g/mL | |
90°C | |
98% min. (GC) | |
C11H22O2 | |
CH3(CH2)8CO2CH3 | |
25 g | |
methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
methyl decanoate | |
8050 | |
98+% |
110-42-9 | |
Colorless | |
226°C | |
Authentic | |
Glass bottle | |
1.4240 to 1.4270 (20°C, 589nm) | |
MFCD00009580 | |
0.871 | |
Solubility in water: practically insoluble | |
CCCCCCCCCC(=O)OC | |
186.30 | |
186.29 | |
Liquid |
Chemical Identifiers
110-42-9 | |
186.30 | |
YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
8050 | |
CCCCCCCCCC(=O)OC |
C11H22O2 | |
MFCD00009580 | |
methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
methyl decanoate |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Warning
EINECSNumber : 203-766-6
RUO – Research Use Only