missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl butyrylacetate, 98+%, Thermo Scientific Chemicals
$60.00 - $60.00
Chemical Identifiers
CAS | 30414-54-1 |
---|---|
Molecular Formula | C7H12O3 |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD00040995 |
InChI Key | SJPCQNABHNCLPB-UHFFFAOYSA-N |
Synonym | methyl butyrylacetate, methyl 3-oxocaproate, 3-oxohexanoic acid methyl ester, hexanoic acid, 3-oxo-, methyl ester, methyl 3-ketohexanoate, butyrylacetic acid methyl ester, 3-oxo-hexanoic acid methyl ester, bem, acmc-1ahmy, ksc497e5t |
PubChem CID | 121700 |
IUPAC Name | methyl 3-oxohexanoate |
SMILES | CCCC(=O)CC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0982614
|
Thermo Scientific Chemicals
L0982614 |
25 g |
Each for $60.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
30414-54-1 | |
144.17 | |
SJPCQNABHNCLPB-UHFFFAOYSA-N | |
121700 | |
CCCC(=O)CC(=O)OC |
C7H12O3 | |
MFCD00040995 | |
methyl butyrylacetate, methyl 3-oxocaproate, 3-oxohexanoic acid methyl ester, hexanoic acid, 3-oxo-, methyl ester, methyl 3-ketohexanoate, butyrylacetic acid methyl ester, 3-oxo-hexanoic acid methyl ester, bem, acmc-1ahmy, ksc497e5t | |
methyl 3-oxohexanoate |
Specifications
30414-54-1 | |
1.037 | |
90°C (194°F) | |
1.426 | |
25 g | |
methyl butyrylacetate, methyl 3-oxocaproate, 3-oxohexanoic acid methyl ester, hexanoic acid, 3-oxo-, methyl ester, methyl 3-ketohexanoate, butyrylacetic acid methyl ester, 3-oxo-hexanoic acid methyl ester, bem, acmc-1ahmy, ksc497e5t | |
CCCC(=O)CC(=O)OC | |
144.17 | |
144.17 | |
Methyl butyrylacetate |
-43°C | |
85°C to 90°C (5 mmHg) | |
C7H12O3 | |
MFCD00040995 | |
636165 | |
SJPCQNABHNCLPB-UHFFFAOYSA-N | |
methyl 3-oxohexanoate | |
121700 | |
≥98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 250-185-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only