Please login to your online account to display your discounted pricing

Methyl butyrate, 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  414142500

 View more versions of this product

Catalog No. AC414142500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 102.13
Physical Form Liquid
Quantity 250g
Color Clear colorless to very slightly yellow
Merck Index 15, 6109
TSCA TSCA
RTECS Number ET5500000
EINECS Number 210-792-1
Density 0.8970g/mL
Packaging Glass bottle
Acid Value 1mg KOH/g max.
Infrared Spectrum Authentic
Refractive Index 1.3850 to 1.3900 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Linear Formula CH3(CH2)2COOCH3
CAS 623-42-7
Assay Percent Range 99%
Chemical Name or Material Methyl butyrate
Name Note 99%
Molecular Formula C5H10O2
MDL Number MFCD00009391

Specifications

Chemical Name or Material Methyl butyrate
Name Note 99%
CAS 623-42-7
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H10O2
Linear Formula CH3(CH2)2COOCH3
Formula Weight 102.13
MDL Number MFCD00009391
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents