missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl α-bromophenylacetate, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 379300050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl α-bromophenylacetate | |
1.4550g/mL | |
113°C | |
Glass bottle | |
C6H5CH(Br)CO2CH3 | |
5g | |
methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate | |
NHFBYYMNJUMVOT-UHFFFAOYSA-N | |
methyl 2-bromo-2-phenylacetate | |
137806 | |
97+% |
3042-81-7 | |
113.0°C (3.0 mmHg) | |
97% min. (GC) | |
C9H9BrO2 | |
MFCD00013535 | |
1.455 | |
Solubility in water: .. Other solubilities: soluble in most organic solvents | |
COC(=O)C(C1=CC=CC=C1)Br | |
229.07 | |
229.07 |
Chemical Identifiers
3042-81-7 | |
229.07 | |
NHFBYYMNJUMVOT-UHFFFAOYSA-N | |
137806 | |
COC(=O)C(C1=CC=CC=C1)Br |
C9H9BrO2 | |
MFCD00013535 | |
methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate | |
methyl 2-bromo-2-phenylacetate |
Safety and Handling
GHS Signal Word: Danger
RUO â Research Use Only