missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl beta-D-xylopyranoside, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6672714

 View more versions of this product

Catalog No. AAJ6672714



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl beta-D-xylopyranoside
155°C to 158°C
C6H12O5
25 g
ZBDGHWFPLXXWRD-JGWLITMVSA-N
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
11768891
164.16
Powder
612-05-5
White
MFCD00047532
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
COC1C(C(C(CO1)O)O)O
164.157
CHEBI:74863
98%

Chemical Identifiers

612-05-5
164.157
ZBDGHWFPLXXWRD-JGWLITMVSA-N
11768891
(2R,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
C6H12O5
MFCD00047532
methyl-beta-d-xylopyranoside, methyl beta-d-xyloside, methyl beta-d-xylopyranoside, beta-methyl-d-xyloside, 2r,3r,4s,5r-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 1-o-methyl-beta-d-xylopyranose, 2r,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-beta-d-xylopyranoside, beta-methylxyloside, beta-d-xylp-ome
CHEBI:74863
COC1C(C(C(CO1)O)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 210-289-7

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only