Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl beta-D-glucopyranoside hemihydrate, 98%

$36.80 - $101.45

Chemical Identifiers

Chemical Name or Material Methyl beta-D-glucopyranoside hemihydrate
CAS 7000-27-3
Molecular Formula C7H14O6·0.5H2O
Formula Weight 203.19 (194.18 Anhydrous)
MDL Number MFCD00149442
Beilstein 4937464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0702603

 
alfa aesar™
L0702603
1g Each for $36.80

AAL0702606

 
alfa aesar™
L0702606
5g Each for $101.45
Description & Specifications

Specifications

Formula Weight 203.19 (194.18 Anhydrous)
Assay 98%
Chemical Name or Material Methyl beta-D-glucopyranoside hemihydrate
TSCA TSCA
MDL Number MFCD00149442
CAS 7000-27-3
Beilstein 4937464
Melting Point 106° to 111°C
Quantity 1g
Optical Rotation −33° (c=1 in Water)
Recommended Storage 2° to 8°C
Molecular Formula C7H14O6·0.5H2O
Safety and Handling

2° to 8°C