missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl beta-D-galactopyranoside, Thermo Scientific Chemicals
$71.86 - $276.16
Chemical Identifiers
CAS | 1824-94-8 |
---|---|
Molecular Formula | C7H14O6 |
Molecular Weight (g/mol) | 194.18 |
MDL Number | MFCD00064357 |
InChI Key | HOVAGTYPODGVJG-VOQCIKJUSA-N |
Synonym | methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose |
PubChem CID | 94214 |
ChEBI | CHEBI:17540 |
IUPAC Name | (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
SMILES | CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0812703
|
Thermo Scientific Chemicals
L0812703 |
1 g |
Each for $71.86
|
|
|||||
AAL0812706
|
Thermo Scientific Chemicals
L0812706 |
5 g |
Each for $276.16
|
|
|||||
Description
It is a weak substrate and effective inducer of beta-D-galactosidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a weak substrate and effective inducer of beta-D-galactosidase.
Solubility
Soluble in water and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
Chemical Identifiers
1824-94-8 | |
194.18 | |
HOVAGTYPODGVJG-VOQCIKJUSA-N | |
94214 | |
(2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
C7H14O6 | |
MFCD00064357 | |
methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose | |
CHEBI:17540 | |
CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Specifications
1824-94-8 | |
C7H14O6 | |
1 g | |
methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose | |
HOVAGTYPODGVJG-VOQCIKJUSA-N | |
CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
194.18 | |
CHEBI:17540 | |
Methyl beta-D-galactopyranoside |
175°C to 179°C | |
MFCD00064357 | |
81569 | |
Soluble in water and methanol. | |
−16° (c=1.5 in Methanol) | |
(2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
94214 | |
194.18 |
Safety and Handling
EINECSNumber : 217-361-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only