Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl-beta-cyclodextrin


Quantity 5g

Manufacturer: alfa aesar™  J6684706

 View more versions of this product

Catalog No. AAJ6684706

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point 180° to 182°C
Chemical Name or Material Methyl-beta-cyclodextrin
CAS 128446-36-6
MDL Number MFCD00074980
Physical Form Powder
Solubility Information Soluble in water
Color White to off-white
TSCA TSCA
Quantity 5g

Specifications

Chemical Name or Material Methyl-beta-cyclodextrin
CAS 128446-36-6
MDL Number MFCD00074980

Applications
Cavity size is the major determinant as to which cyclodextrin is used in complexation. The cavity diameter of β-cyclodextrins or β-glucopyranose unit compounds is well-suited for use with molecules the size of hormones, vitamins and many compounds frequently used in tissue and cell culture applications. For this reason, β-cyclodextrin is most commonly used as a complexing agent. It is used to increase the solubility of non-polar substances such as fatty acids, lipids, vitamins and cholesterol for use in cell culture applications. Methyl-β-cyclodextrin is used to remove cholesterol from cultured cells and is employed to remove lipid rafts.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents.