Please login to your online account to display your discounted pricing

Methyl Benzoylformate 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl benzoylformate
Name Note 99%
CAS 15206-55-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8O3
Linear Formula C6H5COCO2CH3
Formula Weight 164.16
MDL Number MFCD00008443
Synonym Methyl phenylglyoxylate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126370050

  SDS
thermo fisher scientific
AC126370050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 164.16
Physical Form Liquid
Quantity 5g
Color Clear yellow
TSCA TSCA
EINECS Number 239-263-3
Density 1.1500g/mL
Beilstein 10, 657
Refractive Index 1.5250 to 1.5270 (20°C, 589nm)
Synonym Methyl phenylglyoxylate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
CAS 15206-55-0
Assay Percent Range 99%
Chemical Name or Material Methyl benzoylformate
MDL Number MFCD00008443
Molecular Formula C9H8O3
Linear Formula C6H5COCO2CH3