missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (Methyl benzoate)tricarbonylchromium, 98%

Catalyst for hydrogenation of unsaturated fats

Manufacturer:  Alfa Aesar™ 4436803

 View more versions of this product

Catalog No. AA4436803



Specifications

Specifications

(Methyl benzoate)tricarbonylchromium
Orange
1g
C11H8CrO5
UN3466
GEALMZJLYGNHRA-UHFFFAOYSA-N
carbon monoxide;chromium;methyl benzoate
11780645
Crystalline
Air and moisture sensitive
95°C to 96°C
12125-87-0
MFCD00009957
methyl benzoate tricarbonylchromium
COC(=O)C1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]
272.176
272.18
98%

Chemical Identifiers

12125-87-0
272.176
GEALMZJLYGNHRA-UHFFFAOYSA-N
11780645
COC(=O)C1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]
C11H8CrO5
MFCD00009957
methyl benzoate tricarbonylchromium
carbon monoxide;chromium;methyl benzoate
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents