Please login to your online account to display your discounted pricing

Alfa Aesar™ (Methyl benzoate)tricarbonylchromium, 98%

Catalyst for hydrogenation of unsaturated fats

$106.99 - $106.99

Chemical Identifiers

Chemical Name or Material (Methyl benzoate)tricarbonylchromium
CAS 12125-87-0
Molecular Formula C11H8CrO5
Formula Weight 272.18
MDL Number MFCD00009957
UN Number UN3466
Synonym (Methyl benzoate)chromium tricarbonyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4436803

  SDS
alfa aesar™
4436803
1g Each for $106.99

AA4436806

  SDS
alfa aesar™
4436806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 272.18
Health Hazard 1 Danger
Melting Point 95° to 96°C
Chemical Name or Material (Methyl benzoate)tricarbonylchromium
Quantity 1g
Assay 98%
UN Number UN3466
Synonym (Methyl benzoate)chromium tricarbonyl
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Molecular Formula C11H8CrO5
MDL Number MFCD00009957
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Crystalline
CAS 12125-87-0
Color Orange
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.