missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl benzoate, 99%

Manufacturer:  Alfa Aesar A1240430

 View more versions of this product

Catalog No. AAA1240430


 

Description

Description

Applications
Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Methyl Benzoate
250g
99%
C8H8O2
1072099
QPJVMBTYPHYUOC-UHFFFAOYSA-N
methyl benzoate
7150
136.15
1.092
93-58-3
99%
MFCD00008421
14,6024
COC(=O)C1=CC=CC=C1
136.15
CHEBI:72775
Liquid

Chemical Identifiers

93-58-3
136.15
QPJVMBTYPHYUOC-UHFFFAOYSA-N
CHEBI:72775
C8H8O2
MFCD00008421
7150
Safety and Handling

Safety and Handling

GHS H Statement
H302-H227

GHS P Statement
P210-P280-P264-P270-P301+P312-P330-P370+P378a-P403+P235-P501a

Warning

RTECSNumber : DH3850000

TSCA : TSCA

SDS Documents