Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl benzo[b]thiophene-2-carboxylate, 98%

$51.68 - $51.68

Chemical Identifiers

Chemical Name or Material Methyl benzo[b]thiophene-2-carboxylate
CAS 22913-24-2
Molecular Formula C10H8O2S
Formula Weight 192.24
MDL Number MFCD00067791
Beilstein 142166
Synonym Benzo[b]thiophene-2-carboxylic acid methyl ester; Methyl thianaphthene-2-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1376903

  SDS
alfa aesar™
A1376903
1g Each for $51.68
Description & Specifications

Specifications

Formula Weight 192.24
Molecular Formula C10H8O2S
Assay 98%
Quantity 1g
MDL Number MFCD00067791
CAS 22913-24-2
Beilstein 142166
Melting Point 69° to 70°C
Chemical Name or Material Methyl benzo[b]thiophene-2-carboxylate
Synonym Benzo[b]thiophene-2-carboxylic acid methyl ester; Methyl thianaphthene-2-carboxylate