Please login to your online account to display your discounted pricing

Methyl benzimidate hydrochloride, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC205270010

 View more versions of this product

Catalog No. AC205270010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 171.63
Quantity 1g
Color White
Beilstein 09, III, 1250
Synonym Methyl benzenecarboximidate
Health Hazard 1 Warning
CAS 5873-90-5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula C6H5C(=NH)OCH3·HCl
MDL Number MFCD00012575
Assay Percent Range 98%
Molecular Formula C8H9NO·HCl
Chemical Name or Material Methyl benzimidate hydrochloride

Specifications

Chemical Name or Material Methyl benzimidate hydrochloride
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9NO·HCl
Linear Formula C6H5C(=NH)OCH3·HCl
Formula Weight 171.63
MDL Number MFCD00012575
Synonym Methyl benzenecarboximidate
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents