Please login to your online account to display your discounted pricing

Methyl benzimidate hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl benzimidate hydrochloride
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9NO·HCl
Linear Formula C6H5C(=NH)OCH3·HCl
Formula Weight 171.63
MDL Number MFCD00012575
Synonym Methyl benzenecarboximidate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205270010

  SDS
fisher scientific acros
AC205270010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 171.63
Quantity 1g
Color White
Beilstein 09, III, 1250
Chemical Name or Material Methyl benzimidate hydrochloride
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C8H9NO·HCl
Linear Formula C6H5C(=NH)OCH3·HCl
MDL Number MFCD00012575
CAS 5873-90-5
Synonym Methyl benzenecarboximidate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.