missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl benzenesulfonate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 413990250

 View more versions of this product

Catalog No. AC413990250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl benzenesulfonate
80-18-2
Colorless to Yellow
280.0°C
Authentic
Glass bottle
1.5140 to 1.5160
25 g
1.3
CZXGXYBOQYQXQD-UHFFFAOYSA-N
methyl benzenesulfonate
6630
99%
99%
-4.0°C
1.3000g/mL
143°C
98.5% min. (HPLC)
C7H8O3S
MFCD00014737
07,645
benzenesulfonic acid, methyl ester, methyl benzenesulphonate, methyl besylate, methylbenzenesulfonate, ccris 9156, benzenesulfonic acid methyl ester, methyl benzene sulfonate, methyl ester of benzenesulphonic acid, methylbenzene sulfonate, pubchem22730
COS(=O)(=O)C1=CC=CC=C1
172.20
172.2
Liquid

Chemical Identifiers

80-18-2
172.20
CZXGXYBOQYQXQD-UHFFFAOYSA-N
6630
COS(=O)(=O)C1=CC=CC=C1
C7H8O3S
MFCD00014737
benzenesulfonic acid, methyl ester, methyl benzenesulphonate, methyl besylate, methylbenzenesulfonate, ccris 9156, benzenesulfonic acid methyl ester, methyl benzene sulfonate, methyl ester of benzenesulphonic acid, methylbenzene sulfonate, pubchem22730
methyl benzenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 201-256-8

RTECSNumber : DB7151000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only