missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl benzenesulfonate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC413995000

 View more versions of this product

Catalog No. AC413995000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Methyl benzenesulfonate
Clear colorless to slightly yellow
99%
Authentic
Glass bottle
≥98.5% (HPLC)
C7H8O3S
MFCD00014737
1.3000g/mL
500g
80-18-2
1.5150 to 1.5180 (20°C, 589nm)
Liquid
99%
172.2

Chemical Identifiers

80-18-2
C7H8O3S
MFCD00014737
≥98.5% (HPLC)
99%
172.2
Methyl benzenesulfonate, 99%

Safety and Handling

SDS & Safety Documents

GHS P Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-256-8

RTECSNumber : DB7151000

TSCA : TSCA