missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Benzenesulfinate, TCI America™

$111.53 - $334.59

Chemical Identifiers

CAS 670-98-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD00674051
InChI Key PSNSVDSRLUYDKF-UHFFFAOYSA-N
Synonym methylbenzenesulfinate, benzenesulfinic acid methyl ester, methyl phenylsulfinate, methyl benzene sulfinate, benzenesulfinic acid,methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, methoxy phenylsulfinyl, acmc-20mem2, acmc-20mg8f
PubChem CID 2759784
IUPAC Name methyl benzenesulfinate
SMILES COS(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29715G View Documents TCI America
M29715G
5g Each for $111.53
 
M297125G View Documents TCI America
M297125G
25g Each for $334.59
 
Specifications

Chemical Identifiers

670-98-4
156.199
PSNSVDSRLUYDKF-UHFFFAOYSA-N
2759784
COS(=O)C1=CC=CC=C1
C7H8O2S
MFCD00674051
methylbenzenesulfinate, benzenesulfinic acid methyl ester, methyl phenylsulfinate, methyl benzene sulfinate, benzenesulfinic acid,methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, methoxy phenylsulfinyl, acmc-20mem2, acmc-20mg8f
methyl benzenesulfinate

Specifications

63°C
5g
C7H8O2S
methylbenzenesulfinate, benzenesulfinic acid methyl ester, methyl phenylsulfinate, methyl benzene sulfinate, benzenesulfinic acid,methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, methoxy phenylsulfinyl, acmc-20mem2, acmc-20mg8f
COS(=O)C1=CC=CC=C1
156.199
156.20
≥98.0% (GC)
Yellow
670-98-4
MFCD00674051
PSNSVDSRLUYDKF-UHFFFAOYSA-N
methyl benzenesulfinate
2759784
Clear Liquid at 20°C
Methyl Benzenesulfinate
Safety and Handling

Safety and Handling

TSCA : No