missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl benzenesulfinate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 670-98-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.2
InChI Key PSNSVDSRLUYDKF-UHFFFAOYSA-N
Synonym acmc-20mem2, acmc-20mg8f, benzenesulfinic acid methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, benzenesulfinic acid,methyl ester, methoxy phenylsulfinyl, methyl benzene sulfinate, methyl phenylsulfinate, methylbenzenesulfinate
PubChem CID 2759784
IUPAC Name methyl benzenesulfinate
SMILES COS(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC438610050
SDS View Documents Promotion Details
Thermo Fisher Scientific
438610050
5mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC438610250
SDS View Documents Promotion Details
Thermo Fisher Scientific
438610250
25mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

670-98-4
156.2
acmc-20mem2, acmc-20mg8f, benzenesulfinic acid methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, benzenesulfinic acid,methyl ester, methoxy phenylsulfinyl, methyl benzene sulfinate, methyl phenylsulfinate, methylbenzenesulfinate
methyl benzenesulfinate
C7H8O2S
PSNSVDSRLUYDKF-UHFFFAOYSA-N
2759784
COS(=O)C1=CC=CC=C1

Specifications

670-98-4
Colorless to Yellow
97.5% min. (GC)
Authentic
5mL
1.194
COS(=O)C1=CC=CC=C1
156.2
156.2
98%
1.1940g/mL
110.0°C to 112.0°C (16.0 Torr)
C7H8O2S
Glass bottle
acmc-20mem2, acmc-20mg8f, benzenesulfinic acid methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, benzenesulfinic acid,methyl ester, methoxy phenylsulfinyl, methyl benzene sulfinate, methyl phenylsulfinate, methylbenzenesulfinate
PSNSVDSRLUYDKF-UHFFFAOYSA-N
methyl benzenesulfinate
2759784
Liquid
Methyl benzenesulfinate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call

GHS Signal Word: Warning

SDS
Product Certifications

RUO – Research Use Only