Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl arachidonyl fluorophosphonate, 98%

Chemical Identifiers

Chemical Name or Material Methyl arachidonyl fluorophosphonate
CAS 188404-10-6
Formula Weight 370.49
MDL Number MFCD03095722
Synonym MAFP; Methylphosphonofluoridic acid 5,8,11,14-eicosatetraenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67139LB0

  SDS
alfa aesar™
J67139LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 370.49
Chemical Name or Material Methyl arachidonyl fluorophosphonate
Physical Form Oil
MDL Number MFCD03095722
CAS 188404-10-6
Synonym MAFP; Methylphosphonofluoridic acid 5,8,11,14-eicosatetraenyl ester
Quantity 5mg
Sensitivity Air Sensitive
RTECS Number TA8108000
Assay 98%
Color Colorless to yellow
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C