missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl arachidate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 452820050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl Arachidate | |
45.0°C to 48.0°C | |
>110°C | |
C21H42O2 | |
MFCD00009014 | |
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate | |
CCCCCCCCCCCCCCCCCCCC(=O)OC | |
326.57 | |
326.56 |
1120-28-1 | |
215.0°C to 216.0°C (10.0 mmHg) | |
98.5% min. (GC) | |
CH3(CH2)18COOCH3 | |
5 g | |
QGBRLVONZXHAKJ-UHFFFAOYSA-N | |
methyl icosanoate | |
14259 | |
99% |
Chemical Identifiers
1120-28-1 | |
326.57 | |
QGBRLVONZXHAKJ-UHFFFAOYSA-N | |
14259 | |
CCCCCCCCCCCCCCCCCCCC(=O)OC |
C21H42O2 | |
MFCD00009014 | |
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate | |
methyl icosanoate |
Safety and Handling
EINECSNumber : 214-304-8
RUO – Research Use Only